Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ KIMAX™ KimCote™ Heavy Wall, Round Bottom Flask
SDP

Catalog No. 01183001 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
01-183-001 250 mL
01-183-000 100 mL
12-141-339 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 01-183-001 Supplier DWK Life Sciences Supplier No. KC25330250

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Kimble™ KIMAX™ KimCote™ Heavy Wall, Round Bottom Flask is a plastic-coated single neck flask useful as a rotary evaporator condensate collection flask or as a replacement for European designation KS 20/35.

  • Spherical socket joint
  • Joint Size: 35/20 mm
  • Autoclavable KimCote safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
Material Borosilicate Glass (3.3)
Standard Taper Joint 35/25
Autoclavable Autoclavable
Sterility Non-sterile
Bottom Shape Round
Diameter (Metric) Outer 83 mm
Surface Treatment Plastic Coated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.