Learn More
DWK Life Sciences KIMBLE™ KONTES™ MIDI-VAP 4000 Distillation Tubing Kit
SDP
Catalog No. K4794180046 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
MIDI-VAP 4000
MIDI-VAP 4000 Ammonia/Phenol System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
K4794180046 MIDI-VAP 4000
K4794180043 MIDI-VAP 4000 Ammonia/Phenol System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K4794180046 Supplier DWK Life Sciences Supplier No. 4794180046
Add to Cart
Add to Cart
Genuine replacement and optional components for the MIDI-VAP 4000 Distillation Apparatus, supporting ammonia, phenol, and cyanide distillation workflows.
Component parts for both ammonia and phenol distillation system
  • Genuine replacement and optional parts for the Kimble™ Kontes™ MIDI-VAP 4000 Distillation Apparatus
  • Supports ammonia, phenol, and cyanide distillation applications
  • Includes glassware sets, tubing kits, reaction tubes, distilling heads, caps, septa, sealing rings, quick disconnects, and manifold heater
  • Borosilicate glass components designed for reduced sample volume semi-automatic colorimetry methods
  • Compatible with drinking, ground, surface, and saline water analysis as well as domestic and industrial waste testing
TRUSTED_SUSTAINABILITY
Specifications
Includes All Fittings
For Use With (Application) Distillation Systems
Type Cyanide Distillation Tubing Kit
For Use With (Equipment) MIDI-VAP 4000