Learn More
DWK LIFE SCIENCES™ KIMBLE™ KONTES™ One Piece Aluminum Seal for Vials
SDP
Catalog No. 0334078BB Shop All DWK Life Sciences Products
Change view
Click to view available options
Product Type:
Center Disc Tear-out Cap
Complete Tear-off Cap
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
03-340-78BB Center Disc Tear-out Cap
03-340-245 Complete Tear-off Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-340-78BB Supplier DWK LIFE SCIENCES™ Supplier No. 7382013
Add to Cart
Add to Cart
KIMBLE™ KONTES™ One Piece Aluminum Seal for Vials provides secure, reliable closure for vials and bottles. Available in tear-off and tear-out styles, these unlined one-piece aluminum seals offer easy access to septa and vial contents. Fits GPI aluminum finish seal 13.
  • Available in tear-off and tear-out styles for flexible access to vial contents
  • One piece, unlined construction
  • Tear-out style: center disc tears out, leaving the outside edge firmly crimped on the container
  • Tear-off style: seal can be completely removed from vial or bottle
  • Fits GPI aluminum finish seal 13
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Diameter (Metric) Outer 13 mm
Material Aluminum
Pre-slit No
Sterility Non-sterile
Septum None
Quantity 1000/Cs.
Closure Size 13 mm
Autoclavable Not Autoclavable
Product Type Center Disc Tear-out Cap