Learn More
DWK Life Sciences Kimble™ Kontes™ Reducing/Enlarging Connecting Adapters
SDP
Catalog No. k1501000265 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No.:
29/42 Inner Bottom, 24/40 Outer Top
Catalog No. Standard Taper Joint No.
K150100-0265 29/42 Inner Bottom, 24/40 Outer Top
1 options
Catalog No. K150100-0265 Supplier DWK Life Sciences Supplier No. 1501000265
Only null left
Add to Cart
Add to Cart
For enlarging the size of Standard Taper glass joints.
  • Outer joint is at the top, inner at the bottom
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type 1 and ASTM E438, Type 1, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
For Use With (Application) For connecting dissimilar joints
Material Borosilicate Glass
Product Type Enlarging Connecting Adapter
Standard Taper Joint No. 29/42 Inner Bottom, 24/40 Outer Top