Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Linear HDPE Stoppers
SDP

Catalog No. 146441B Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
9
13
16
19
22
27
Stopper Number:
9
13
16
19
22
27
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Stopper Number
14-644-1B 9 9
14-644-1E 19 19
14-644-1G 27 27
14-644-1C 13 13
14-644-1F 22 22
14-644-1D 16 16
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-644-1B Supplier DWK Life Sciences Supplier No. 28160R9
Only null left
Add to Cart
Add to Cart

Closed bottom end fits the standard taper neck

  • Heavy-duty Polyethylene stopper
  • Flange on stopper protects glass

TRUSTED_SUSTAINABILITY

Specifications

Product Type Tapered Stopper
Stopper Number 9
Size 9
Material HDPE
Color Yellow, Blue
For Use With (Application) Used as replacement parts for items that are ground to standard taper stopper dimensions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.