Learn More
DWK LIFE SCIENCES™ KIMBLE™ Micro-Hematocrit Capillary Tube
SDP
Catalog No. 12141536 Shop All DWK Life Sciences Products
Change view
Click to view available options
Product Type:
Microhematocrit Heparinized Capillary Tube
Catalog No. Product Type
12-141-536 Microhematocrit Heparinized Capillary Tube
1 options
This item is not returnable. View return policy
Catalog No. 12-141-536 Supplier DWK LIFE SCIENCES™ Supplier No. 40A502
Add to Cart
Add to Cart
This item is not returnable. View return policy
KIMBLE™ Micro-Hematocrit Capillary Tube is designed to prevent blood clotting. Packed in plastic vials with reclosable snap-lock caps, these soda lime glass tubes conform to USP Type III requirements and can be sealed by flame or sealing clay.
  • Heparinized capillary tubes are designed to prevent blood clotting
  • Packed in plastic vials with reclosable snap-lock caps to maintain cleanliness
  • Ends may be sealed by flame or sealing clay
  • Ammonium heparin does not interfere with sodium identification
  • 44B508 and 44A509 are bulk quantities
  • Tubes are manufactured from 90 expansion soda lime glass conforming to USP Type III requirements
  • Available in borosilicate glass as a special order
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Product Type Microhematocrit Heparinized Capillary Tube
Additive Heparin
Material Soda Lime Glass
Sterility Non-sterile
Closure Type Capillary Sealing Compound
Capacity (Metric) 70 μL
Length (Metric) 1.1 x 75 mm