Learn More
DWK Life Sciences™ Kimble™ Nickel-Plated, Standard Taper Joint Clamp
SDP
Catalog No. 01183044 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint:
14/20, 14/35
19/22, 19/38
24/40, 24/25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
01-183-044 24/40, 24/25
01-183-042 14/20, 14/35
01-183-043 19/22, 19/38
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 01-183-044 Supplier DWK Life Sciences™ Supplier No. 6753200024
Add to Cart
Add to Cart
This item is not returnable. View return policy
This Kimble™ Nickel-Plated metal, Standard Taper Joint Clamp designed to secure Standard Taper joints. The nickel-plating provides increased resistance to heat and cracking.
  • Manufactured from Nickel-plated metal
  • When using a concentrator tube heating mantle, these clamps may be used to secure the concentrator tube to the concentrator flask
  • They can withstand temperatures as high as 200°C, and they have excellent chemical resistance
  • Easy and safe to put on and are dimensionally stable
TRUSTED_SUSTAINABILITY
Specifications
Type Standard Taper Joint Clamp
Material Nickel-Plated Metal
Standard Taper Joint 24/40, 24/25