Learn More
DWK Life Sciences KIMBLE™ Replacement Stopcocks for KIMAX™ Microburet with Three-Way PTFE Stopcock
SDP
Catalog No. 1463153A Shop All DWK Life Sciences Products
Change view
Click to view available options
Plug Bore:
2 mm
Catalog No. Plug Bore
14-631-53A 2 mm
1 options
Catalog No. 14-631-53A Supplier DWK Life Sciences Supplier No. 41530F2
Add to Cart
Add to Cart
KIMBLE™ Replacement Stopcocks for KIMAX™ Microburet with Three-Way PTFE Stopcock are chemically resistant and self-lubricating PTFE plugs designed for reservoir-fill burets and pipets. Each plug is furnished with a nut, rubber washer, and HDPE washer.
  • Replacement three-way PTFE stopcock plug for KIMAX™ microburets and reservoir-fill burets and pipets requiring standard taper PTFE stopcock plugs
  • Chemically resistant and self-lubricating PTFE construction
  • Plug size: 12.9/44 mm
  • Each plug furnished with a nut, rubber washer, and HDPE washer
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement Stopcock
Plug Bore 2 mm
For Use With (Application) Microburet
For Use With (Equipment) Kimax Microburet
Includes Nut, rubber and HPDE washers
Plug Material PTFE
Connectors 3-way