Learn More
DWK Life Sciences™ Kimble™ Stainless Steel Standard Taper Joint Clamp
SDP
Catalog No. 01183045 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint:
14/20, 14/35
Catalog No. Standard Taper Joint
01-183-045 14/20, 14/35
1 options
This item is not returnable. View return policy
Catalog No. 01-183-045 Supplier DWK Life Sciences™ Supplier No. 6754001420
This item is not returnable. View return policy
The Kimble™ Stainless Steel Standard Taper Joint Clamp is used to secure standard ground joint connections in glass assemblies and is characterized by its easy handling and excellent resilience.
  • Available in either metal or polyoxymethylene (POM)
  • Dimensionally stable with particularly good spring properties
  • Easy, safe and quick to fit
  • Superior chemical and temperature resistance
  • Can be heated up to 500°C
  • Plastic clips are marked with their joint size and are color coded to allow easy identification of the size of clip required
TRUSTED_SUSTAINABILITY
Specifications
Type Standard Taper Joint Clamp
Material Stainless Steel
Standard Taper Joint 14/20, 14/35