Learn More
Description
- Gray butyl rubber construction for reliable sealing performance
- Designed to fit GPI aluminum seal finish 20 mm headspace vials
- Provides secure closure to maintain sample integrity
- Compatible with standard headspace vial crimping workflows
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (Metric) Bottom | 13 mm |
| Diameter (Metric) Outer | 20 mm |
| Diameter (Metric) Top | 20 mm |
| Product Type | Serum Bottle Stopper |
| Shape | Cylindrical |
| Sterility | Non-sterile |
| Stopper Number | 21 |
| Size | 20 |
| Color | Gray |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
