Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Straight-Sided Serum Stoppers
SDP

Catalog No. 03340300 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
Chlorobutyl Rubber
Chlorobutyl Rubber, PTFE
Diameter (Metric) Outer:
13 mm
20 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Diameter (Metric) Outer
03-340-300 Chlorobutyl Rubber, PTFE 13 mm
03-340-302 Chlorobutyl Rubber, PTFE 20 mm
03-340-303 Chlorobutyl Rubber 20 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-340-300 Supplier DWK Life Sciences Supplier No. 73811T13
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For research and pharmaceutical packaging applications

  • For use with 13 and 20mm serum-finished bottles and vials
  • Universal gray chlorobutyl formulation meets USP, EP, and JP requirements for globally marketed pharmaceutical products
  • Contain no plasticizers, 2-mercapto-benzothiazole, nitrosamine precursors or natural rubber latex
  • Applicable to aqueous solutions with a pH range of 2 to 10
  • PTFE facing improves chemical compatibility and minimizes surface interactions

Order Info

Durometer 50.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) 13 mm GPI Aluminum Seal
Diameter (Metric) Outer 13 mm
Hardness 50 Durometer
For Use With (Application) For research and pharmaceutical packaging applications
Description Latex free
Color Gray
Material Chlorobutyl Rubber, PTFE
Product Type Plug Stopper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.