Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences KIMBLE™ Straight-Sided Serum Stoppers
SDP

Catalog No. 03340302 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Outer:
13 mm
20 mm
Quantity:
100/Cs.
1000/Cs.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer Quantity
03-340-302 20 mm 100/Cs.
03-340-303 20 mm 1000/Cs.
03-340-300 13 mm 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-340-302 Supplier DWK Life Sciences Supplier No. 73811T21
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KIMBLE™ Straight-Sided Serum Stoppers are designed for research and pharmaceutical packaging applications. The universal gray chlorobutyl formulation meets USP, EP, and JP requirements and is free of plasticizers, nitrosamine precursors, and natural rubber latex.

  • For use with 13mm and 20mm serum-finished bottles and vials
  • Universal gray chlorobutyl formulation meets USP, EP, and JP requirements for globally marketed pharmaceutical products
  • Contain no plasticizers, 2-mercapto-benzothiazole, nitrosamine precursors, or natural rubber latex
  • Applicable to aqueous solutions with a pH range of 2 to 10
  • PTFE facing (73811T series) improves chemical compatibility and minimizes surface interactions
  • Durometer 50

Order Info

Durometer 50.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Bottom 13 mm
Quantity 100/Cs.
Autoclavable Autoclavable
Diameter (Metric) Top 20 mm
Product Type Straight Plug Stopper
Shape Cylindrical
Sterility Non-sterile
Stopper Number 20
Diameter (Metric) Outer 20 mm
For Use With (Equipment) 20 mm GPI Aluminum Seal
Hardness 50 Durometer
Description Latex free
Color Gray
For Use With (Application) For research and pharmaceutical packaging applications
Certifications/Compliance USP, EP, JP
Material Chlorobutyl Rubber, PTFE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.