Learn More
Description
- For use with 13mm and 20mm serum-finished bottles and vials
- Universal gray chlorobutyl formulation meets USP, EP, and JP requirements for globally marketed pharmaceutical products
- Contain no plasticizers, 2-mercapto-benzothiazole, nitrosamine precursors, or natural rubber latex
- Applicable to aqueous solutions with a pH range of 2 to 10
- PTFE facing (73811T series) improves chemical compatibility and minimizes surface interactions
- Durometer 50
Order Info
Durometer 50.
Specifications
Specifications
| Quantity | 1000/Cs. |
| Product Type | Straight Plug Stopper |
| Autoclavable | Autoclavable |
| Diameter (Metric) Bottom | 13 mm |
| Diameter (Metric) Top | 20 mm |
| Material | PTFE-coated Chlorobutyl Rubber |
| Sterility | Non-sterile |
| Stopper Number | 20 |
| Shape | Cylindrical |
| For Use With (Equipment) | 20 mm GPI Aluminum Seal |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
