Learn More
Thermo Scientific™ Kimble Vials For Shimadzu ASI-5000
Catalog No. 0334058 Shop All Thermo Scientific Products
Catalog No. 03-340-58 Supplier Thermo Scientific™ Supplier No. 608381296
Available without closure or preassembled with polypropylene cap and PTFE septa
  • 5mL capacity screw-thread autosampler vial assembly is designed for use with 10-425 GPI polypropylene cap
  • O.D. × L: 12 × 96mm
  • 03-340-58 is similar to 03-340-56C, except without GPI thread finish