Learn More
Research Products International Corp KimCote Plastic Coated Erlenmeyer Flask, 500 ml, 6 per Case
SDP
Catalog No. 501977266
Encompass
KimCote Plastic Coated Erlenmeyer Flasks feature a unique non-slip surface for safe handling in both wet and dry conditions. Heavy-duty plastic coating adds protections from shattering and spilling of contents should a break occur. KimCote is ultra-clear and will not obstruct visibility of contents. Borosilicate flask has white graduations and marking spot. Supplied with a blue polypropylene cap with molded in seal and dripless pour ring.
Catalog No. 50-197-7266 Supplier Research Products International Corp Supplier No. 218221
KimCote Plastic Coated Erlenmeyer Flasks feature a unique non-slip surface for safe handling in both wet and dry conditions. Heavy-duty plastic coating adds protections from shattering and spilling of contents should a break occur. KimCote is ultra-clear and will not obstruct visibility of contents. Borosilicate flask has white graduations and marking spot. Supplied with a blue polypropylene cap with molded in seal and dripless pour ring.