Learn More
MSC Kimtech Disposable Gloves, Sterling Nitrile-Xtra, Nitrile, Powder-Free, Medium, 100/Pack
Catalog No. 5000788250
Encompass
These disposable gloves are part of the Sterling Nitrile-Xtra series, offering general purpose hand protection. Made from nitrile, they are powder-free and feature a grain grip on the fingertips, suitable for various laboratory and industrial applications.
  • Nitrile primary material
  • Has a thickness of 3.5 mil
  • Grain grip on fingertips
  • General purpose grade
  • Regular cuff type
  • Has a length of 9.5 in
  • Medium size
  • Gray color
  • Acceptable quality level (AQL) of 1
  • Non-sterile
  • Powder-free
  • Rolled cuff style
  • Ambidextrous design
  • Uncoated material
Catalog No. 50-007-88250 Supplier MSC Supplier No. 66730334
These disposable gloves are part of the Sterling Nitrile-Xtra series, offering general purpose hand protection. Made from nitrile, they are powder-free and feature a grain grip on the fingertips, suitable for various laboratory and industrial applications.
  • Nitrile primary material
  • Has a thickness of 3.5 mil
  • Grain grip on fingertips
  • General purpose grade
  • Regular cuff type
  • Has a length of 9.5 in
  • Medium size
  • Gray color
  • Acceptable quality level (AQL) of 1
  • Non-sterile
  • Powder-free
  • Rolled cuff style
  • Ambidextrous design
  • Uncoated material