Learn More
MSC Kimtech Nitrile Exam Gloves, Polaris, Large, 5.9 mil, 100/Box
Catalog No. 5000769535
Encompass
The Polaris Nitrile Exam Gloves offer protection, durability, and comfort for scientific applications. Featuring a high-quality nitrile formulation, these gloves ensure high protection, meet and surpass industry standards, and are recyclable through the RightCycle Program.
  • Tested for use with Fentanyl and 24 chemotherapy drugs; meets USP 800 guidelines
  • Certified by US Ergonomics; ambidextrous with textured fingertips for enhanced grip
  • Meets FDA food contact requirements (21 CFR 177.2600)
  • Strong and durable with AQL 0.65, exceeding ASTM AQL 2.5 for critical defects
  • Recyclable via The RightCycle Program; provides protection against harsh chemicals
  • Average tensile strength of 31 Mpa (after aging)
Catalog No. 50-007-69535 Supplier MSC Supplier No. 04203493
The Polaris Nitrile Exam Gloves offer protection, durability, and comfort for scientific applications. Featuring a high-quality nitrile formulation, these gloves ensure high protection, meet and surpass industry standards, and are recyclable through the RightCycle Program.
  • Tested for use with Fentanyl and 24 chemotherapy drugs; meets USP 800 guidelines
  • Certified by US Ergonomics; ambidextrous with textured fingertips for enhanced grip
  • Meets FDA food contact requirements (21 CFR 177.2600)
  • Strong and durable with AQL 0.65, exceeding ASTM AQL 2.5 for critical defects
  • Recyclable via The RightCycle Program; provides protection against harsh chemicals
  • Average tensile strength of 31 Mpa (after aging)