Learn More
MSC Kimtech Nitrile Exam Gloves, Polaris, X-Small, 9.5 in, 100/Bx
Catalog No. 5000795639
Encompass
The Kimtech Polaris Nitrile Exam Gloves are engineered for laboratory environments, offering robust protection against harsh chemicals and chemotherapy drugs. Constructed from a proprietary high-quality nitrile formulation, these gloves feature textured fingertips for superior wet and dry grip, designed for both strength and comfort.
  • Textured fingertips to enhance wet and dry grip
  • Provides protection against harsh chemicals and chemotherapy drugs per ASTM D6978-05
  • Proprietary high-quality nitrile formulation delivers a high level of protection
  • Meets FDA food contact requirements (21 CFR 177.2600)
  • Average tensile strength of 31 Mpa (after aging)
  • Recyclable through the RightCycle™ Program
Catalog No. 50-007-95639 Supplier MSC Supplier No. 04203303
The Kimtech Polaris Nitrile Exam Gloves are engineered for laboratory environments, offering robust protection against harsh chemicals and chemotherapy drugs. Constructed from a proprietary high-quality nitrile formulation, these gloves feature textured fingertips for superior wet and dry grip, designed for both strength and comfort.
  • Textured fingertips to enhance wet and dry grip
  • Provides protection against harsh chemicals and chemotherapy drugs per ASTM D6978-05
  • Proprietary high-quality nitrile formulation delivers a high level of protection
  • Meets FDA food contact requirements (21 CFR 177.2600)
  • Average tensile strength of 31 Mpa (after aging)
  • Recyclable through the RightCycle™ Program