Learn More
MSC Kimberly-Clark Professional Sterile Wipers, Polypropylene, 70% IPA/30% DIW, 11 x 9 in
Catalog No. 502837817
Encompass
These sterile presaturated alcohol wipers are designed for critical sterile environments like cleanrooms and biotech. Made from 100% polypropylene, they are pre-saturated with 70% isopropyl alcohol (semiconductor grade) and 30% deionized water. They are low-lint, suitable for ISO Class 4 and higher cleanrooms, and ideal for tasks in pharmaceutical, high-tech, and medical-device industries.
  • 100% polypropylene, pre-saturated with 70% IPA/30% DIW
  • Thermally bonded; acid-, base-, and solvent-resistant
  • Straight, knife-cut edges for minimal lint
  • One-step alcohol application for critical surfaces
  • Low lint, suitable for ISO Class 4+ cleanrooms
  • Resealable pouch, has an expiration date
Catalog No. 50-283-7817 Supplier MSC Supplier No. 36735447
These sterile presaturated alcohol wipers are designed for critical sterile environments like cleanrooms and biotech. Made from 100% polypropylene, they are pre-saturated with 70% isopropyl alcohol (semiconductor grade) and 30% deionized water. They are low-lint, suitable for ISO Class 4 and higher cleanrooms, and ideal for tasks in pharmaceutical, high-tech, and medical-device industries.
  • 100% polypropylene, pre-saturated with 70% IPA/30% DIW
  • Thermally bonded; acid-, base-, and solvent-resistant
  • Straight, knife-cut edges for minimal lint
  • One-step alcohol application for critical surfaces
  • Low lint, suitable for ISO Class 4+ cleanrooms
  • Resealable pouch, has an expiration date