Learn More
Aviva Systems Biology Corp KIN Antibody - middle region (ARP31690_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31690_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIN. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human KIN. Alternate names: BTCD, KIN17ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-52837