Learn More
Koehler Instrument Company Inc KIN VIS BATH 115-240V 50/60HZ
SDP
Catalog No. NC3452305
Encompass_Preferred
  • Precision-engineered for accurate viscosity measurements.
  • Microprocessor PID/PIO circuitry for reliable temperature control.
  • Accommodates up to 7 capillary viscometers.
  • Temperature range of 20°C to 100°C with 20°C to 105°C option.
  • Temperature display resolution of 0.01°C.
  • Temperature stability of ±0.02°C.
  • Dual digital display for setpoint and actual temperature.
  • Selectable Fahrenheit or Celsius temperature scale.
  • Robust stainless steel construction.
  • Integrated safety features prevent overtemperature and low liquid levels.
  • LED illumination for enhanced visibility.
  • Built-in cooling coil for external temperature control options.
  • Power requirements of 115-240V 50/60Hz.
Catalog No. NC3452305 Supplier Koehler Instrument Company Inc Supplier No. K23900
Add to Cart
Add to Cart
Request a Quote
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
  • Precision-engineered for accurate viscosity measurements.
  • Microprocessor PID/PIO circuitry for reliable temperature control.
  • Accommodates up to 7 capillary viscometers.
  • Temperature range of 20°C to 100°C with 20°C to 105°C option.
  • Temperature display resolution of 0.01°C.
  • Temperature stability of ±0.02°C.
  • Dual digital display for setpoint and actual temperature.
  • Selectable Fahrenheit or Celsius temperature scale.
  • Robust stainless steel construction.
  • Integrated safety features prevent overtemperature and low liquid levels.
  • LED illumination for enhanced visibility.
  • Built-in cooling coil for external temperature control options.
  • Power requirements of 115-240V 50/60Hz.