Learn More
Medchemexpress LLC HY-19961 5mg Medchemexpress, KIN1408 CAS:1903800-11-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY199615MG
Encompass_Preferred
Medchemexpress, HY-19961 5mg KIN1408 CAS:1903800-11-2 KIN1408 is an agonist of the RIG-1-like receptor (RLR) pathway and exhibits a broad-spectrum antiviral activity. KIN1408 exhibits activity against HCV, influenza A, dengue virus 2, Ebola, Nipah, and Lassa viruses. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2201