Learn More
Enzo Life Sciences SCREEN-WELL Kinase Inhibitor library (500 µl/well)
SDP
Catalog No. 502010209
Encompass_Preferred
The SCREEN-WELL Kinase Inhibitor Library contains 80 known kinase inhibitors of well-defined activity. Inhibitors are supplied dissolved in DMSO at 10 mM and aliquoted into deep-well plates at either 100 or 500 microliters per well. The library is an ideal tool for chemical genomics, assay development and other pharmacological applications. Includes inhibitors of these important kinases: Insulin/IGF Receptors, PI 3-Kinase, CaM Kinase II, JAK, PKA, CDK, JNK, PKC, CKI II, MAPK, RAF, EGFR, MEK, SAPK, GSK, MLCK, Src-family, IKK, PDGFR, VEGFR and many more. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0209 Supplier Enzo Life Sciences Supplier No. BML28320500
Add to Cart
Add to Cart
Request a Quote
The SCREEN-WELL Kinase Inhibitor Library contains 80 known kinase inhibitors of well-defined activity. Inhibitors are supplied dissolved in DMSO at 10 mM and aliquoted into deep-well plates at either 100 or 500 microliters per well. The library is an ideal tool for chemical genomics, assay development and other pharmacological applications. Includes inhibitors of these important kinases: Insulin/IGF Receptors, PI 3-Kinase, CaM Kinase II, JAK, PKA, CDK, JNK, PKC, CKI II, MAPK, RAF, EGFR, MEK, SAPK, GSK, MLCK, Src-family, IKK, PDGFR, VEGFR and many more. Long Term Storage: -80°C. Applications: HTS .