Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Kinase Tracer 222, 50 μM

Catalog No. PV6121
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV6121 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV6121 Supplier Thermo Scientific™ Supplier No. PV6121
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Kinase Tracer 222 is an Alexa Fluor™ 647 dye-labeled, ATP-competitive kinase inhibitor scaffold for use in the LanthaScreen™ Eu Kinase Binding Assay.

The LanthaScreen™ Eu Kinase Binding Assay
• Is easily optimized
• Measures inhibitor binding
• Performs simple off-rate measurements

Easily Screen for Kinase Binding
The LanthaScreen™ Eu Kinase Binding Assay is a TR-FRET assay that lets you measure inhibitor binding to non-activated kinases, perform simple off-rate measurements, and makes simple the study of type III (allosteric) inhibitors.

The LanthaScreen™ Eu Kinase Binding Assay is easy to optimize and run. The assay uses a simple mix-and-read format with a single one hour incubation and no development steps. Binding of an Alexa Fluor™ conjugate or “tracer” to a tagged kinase is detected by addition of a europium-labeled anti-tag antibody, in this case anti-GST. Simultaneous binding of the tracer and antibody to the kinase results in a high degree of fluorescence resonance energy transfer (TR-FRET), while displacement of the tracer with a kinase inhibitor results in a loss of TR-FRET.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

TRUSTED_SUSTAINABILITY

Specifications

Format 384-well plate
For Use With (Application) TR-FRET
Quantity 25 μL
Product Type Kinase Tracer 222
Content And Storage Store at -20°C
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Readout Kinetic
Shipping Condition Dry Ice
Target Kinases (Multiple)
Label or Dye Alexa Fluor™ 647
Color Red
Product Line LanthaScreen
Emission 665
Excitation 340
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.