Learn More
Kinematica HMC Europe Vertical Autoclave Air Filters
Catalog No. 08451434 Shop All Kinematica Products
Change view
Click to view available options
For Use With (Equipment):
HMC-HV autoclave
Product Type:
Exhaust Air Filter
Catalog No. For Use With (Equipment) Product Type
08-451-434 HMC-HV autoclave Exhaust Air Filter
1 options
Catalog No. 08-451-434 Supplier Kinematica HMC Europe Supplier No. 50004
Exhaust air filter. Safety for individuals and the environment. Option for safe, emission-free sterilization of highly infectious germs and genetically modified materials. Not for applications requiring FDA approval.
During the venting phase, the steam displaces unsterilized residual air from the chamber and passes it through a filter system. The surrounding air is protected against contamination as the cycle is running. Naturally, in-line sterilization of the exhaust air filter is performed. Due to the design, condensate sterilization is a matter of course. Any condensate which precipitates flows back into the chamber and gets sterilized. Discharge can be done safely.
Specifications
For Use With (Application) Sterilization of exhaust air
For Use With (Equipment) HMC-HV autoclave
Product Type Exhaust Air Filter