Learn More
Kinematica HMC Vertical Autoclave Baskets
Catalog No. 08451433 Shop All Kinematica Products
Change view
Click to view available options
For Use With (Equipment):
HV 85/110 autoclave
Catalog No. For Use With (Equipment)
08-451-433 HV 85/110 autoclave
1 options
Catalog No. 08-451-433 Supplier Kinematica Supplier No. 50023
With this item you can easily and safely bring your sterilized goods into and out of the autoclave; we offer baskets in various sizes, with and without bowl or closed cans with and without holes.
For use with the HMC-HG and HMC-HV autoclaves.
Specifications
For Use With (Equipment) HV 85/110 autoclave
Material Stainless Steel