Learn More
Kinematica Polytron™ Homogenizer Generator Tool
Catalog No. 08451192 Shop All Kinematica Products
Change view
Click to view available options
For Use With (Equipment):
Polytron™ generators
Catalog No. For Use With (Equipment)
08-451-192 Polytron™ generators
1 options
Catalog No. 08-451-192 Supplier Kinematica Supplier No. 11090010
Add to Cart
Add to Cart
Designed for quick and easy service of Polytron™ generators.
  • Fits all Polytron™ generators between 5 and 60 mm
  • Precision made by skilled Swiss craftsmen
Specifications
Description Homogenizer tool
For Use With (Application) Changing rotors on homogenizers
For Use With (Equipment) Polytron™ generators