Learn More
Kinematica Polytron™ 2500E Homogenizer
Catalog No. 08451323 Shop All Kinematica Products
Change view
Click to view available options
Voltage:
230 V
Catalog No. Voltage
08-451-323 230 V
1 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 08-451-323 Supplier Kinematica Supplier No. 11010055
Add to Cart
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Includes
  • upport stand
Requires
Generators (sold separately; see Polytron Accessories)
Versatile units combine quality, convenience and high performance for rapid, reproducible sample processing and consistent sample-to-sample uniformity
  • Variable speed control
  • For heavy-duty use
  • High-speed, high-performance motor
  • Stand included
  • Soft-start motor prevents splashing and sample loss
  • Quick-connect: change generator with one hand
  • Accept wide range of stainless-steel Polytron generators (order separately)
Pulverizing plant and animal tissues, grains and seeds; soil extractions; DNA/RNA extractions, Karl Fischer sample prep, toxicology and food science
Specifications
Voltage 230 V
Model PT 2500 E