Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer DCT-08 Manual Decapping Tool, 8 mm Dia

Catalog No. NC1991367 Shop All Cole-Parmer Products
Encompass_Preferred

Kinesis DCT-08 Decapping Tool, 8mm dia

Catalog No. NC1991367 Supplier Cole-Parmer Supplier No. 9870684
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kinesis DCT-08 Decapping Tool, 8mm dia

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.