Learn More
Cole-Parmer Disposable Syringes, Luer-Lok, 20 mL; 100/Pk
Catalog No. NC1466115 Shop All Cole-Parmer Products
Encompass_Preferred
The two-part, all-plastic syringes are compatible with a wide range of chemicals. The barrel and plunger are made of inert, chemical-resistant polypropylene (PP). Transparent barrels and permanent graduations simplify the monitoring and inspection of liquids. Syringes are nonsterile and have a standard conical fitting with 6% (Luer) taper. These disposable syringes are available with two different connector types: Luer-Lok or Luer-slip. Luer-Lok syringes have a twist-and-lock mechanism to create a secure fitting connection, making them a safe choice for high back pressure liquids in filter applications. Luer-slip syringes slide into place and are held by frictionideal for low-pressure applications.
Catalog No. NC1466115 Supplier Cole-Parmer Supplier No. 1291503
The two-part, all-plastic syringes are compatible with a wide range of chemicals. The barrel and plunger are made of inert, chemical-resistant polypropylene (PP). Transparent barrels and permanent graduations simplify the monitoring and inspection of liquids. Syringes are nonsterile and have a standard conical fitting with 6% (Luer) taper. These disposable syringes are available with two different connector types: Luer-Lok or Luer-slip. Luer-Lok syringes have a twist-and-lock mechanism to create a secure fitting connection, making them a safe choice for high back pressure liquids in filter applications. Luer-slip syringes slide into place and are held by frictionideal for low-pressure applications.