Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 100 x 3.0 mm

Supplier:  Phenomenex Inc 00D4500Y0

Encompass

Kinetex 1.7 m Phenyl-Hexyl 100 00D-4500-Y0

Catalog No. 50-239-5083


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.