Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Kinetex 5 µm C18 100 Å

Catalog No. 502395672 Shop All Phenomenex Products
Encompass

Kinetex 5 µm C18 100 Å Bulk Packing, 1 g

Catalog No. 50-239-5672 Supplier Phenomenex Inc Supplier No. 04A4601
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kinetex 5 µm C18 100 Å Bulk Packing, 1 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.