Learn More
Lonza Walkersville Kinetic-QCL Kinetic Chromogenic LAL Assay, 192 Test Kit
Catalog No. NC9597521
Encompass
The Kinetic-QCL Kinetic Chromogenic LAL Assay is a quantitative, kinetic assay for the detection of Gram-negative bacterial endotoxin. A sample is mixed with the reconstituted LAL reagent in a 96-well plate and placed in an incubating plate reader that measures absorbance at 405 nm. The reaction is automatically monitored over time for the appearance of a yellow color.Benefits:Most sensitive LAL-based methodLess sensitive to product inhibitionApplications:Ideal for biological products such as vaccines and antibioticsContent:8 x 24 tests/vial LAL lysate2 vials endotoxin3 x 30 mL/vial LAL Reagent Water
Catalog No. NC9597521 Supplier Lonza Walkersville Supplier No. 50650U
The Kinetic-QCL Kinetic Chromogenic LAL Assay is a quantitative, kinetic assay for the detection of Gram-negative bacterial endotoxin. A sample is mixed with the reconstituted LAL reagent in a 96-well plate and placed in an incubating plate reader that measures absorbance at 405 nm. The reaction is automatically monitored over time for the appearance of a yellow color.Benefits:Most sensitive LAL-based methodLess sensitive to product inhibitionApplications:Ideal for biological products such as vaccines and antibioticsContent:8 x 24 tests/vial LAL lysate2 vials endotoxin3 x 30 mL/vial LAL Reagent Water