Learn More
Lonza Walkersville Kinetic-QCL Bulk Kinetic Chromogenic LAL Assay, 2400 Test Kit
Catalog No. NC9934559
Encompass
This is a bulk configuration of the KQCL Kinetic Chromogenic LAL Endotoxin Detection Assay and is available as a made-to-order product. This bulk kit configuration contains endotoxin from E. coli Strain O55:B5. Testing of specific lots for making larger product reserves is possible. Please contact your local sales representative for further details. The Kinetic-QCL Kinetic Chromogenic LAL Assay is a quantitative, kinetic assay for the detection of Gram-negative bacterial endotoxin. A sample is mixed with the reconstituted LAL reagent in a 96-well plate and placed in an incubating plate reader that measures absorbance at 405 nm. The reaction is automatically monitored over time for the appearance of a yellow color.Benefits:Most sensitive LAL-based methodLess sensitive to product inhibitionApplications:Ideal for biological products such as vaccines and antibioticsContent:100 x 24 tests/vial LAL Lysate10 vials endotoxin
Catalog No. NC9934559 Supplier Lonza Walkersville Supplier No. 50650H
Add to Cart
Add to Cart
Request a Quote
This is a bulk configuration of the KQCL Kinetic Chromogenic LAL Endotoxin Detection Assay and is available as a made-to-order product. This bulk kit configuration contains endotoxin from E. coli Strain O55:B5. Testing of specific lots for making larger product reserves is possible. Please contact your local sales representative for further details. The Kinetic-QCL Kinetic Chromogenic LAL Assay is a quantitative, kinetic assay for the detection of Gram-negative bacterial endotoxin. A sample is mixed with the reconstituted LAL reagent in a 96-well plate and placed in an incubating plate reader that measures absorbance at 405 nm. The reaction is automatically monitored over time for the appearance of a yellow color.Benefits:Most sensitive LAL-based methodLess sensitive to product inhibitionApplications:Ideal for biological products such as vaccines and antibioticsContent:100 x 24 tests/vial LAL Lysate10 vials endotoxin