Learn More
Apexbio Technology LLC Kinetin 525-79-1 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A35285.1
Encompass_Preferred
Kinetin (N6-furfuryladenine) is a natural adenine derivative classified as a cytokinin originating from plant tissues and participates in diverse physiological processes including cell division and cellular differentiation Structurally kinetin contains an adenine core with a furfuryl substituent at position N6 Experimentally kinetin induces apoptosis-like morphological changes nuclear fragmentation and hypoploidy in vitro in various cell types Additionally kinetin contributes to modulation of RNA splicing patterns notably demonstrated through the promotion of exon 20 inclusion in familial dysautonomia models Due to these properties kinetin serves as an investigational tool in studies of RNA processing apoptosis mechanisms and is examined as a potential therapeutic agent in splicing-associated diseases
Catalog No. 50-000-63003