Learn More
Bioworld Kinetin, 1 g
Catalog No. 50255055
Encompass_Preferred
Kinetin (6-Furfuryl-aminopurine)Synonym: N6-FurfuryladenineCytokinin growth regulatorPlant tissue-culture testedAppearance: white crystaline powderMelting Point: 269-271°CSoluble in 1N NaOHSolubilizeat 1 mg/mLin 0.1N KOH
Catalog No. 50-255-055 Supplier Bioworld Supplier No. 306310141
Kinetin (6-Furfuryl-aminopurine)Synonym: N6-FurfuryladenineCytokinin growth regulatorPlant tissue-culture testedAppearance: white crystaline powderMelting Point: 269-271°CSoluble in 1N NaOHSolubilizeat 1 mg/mLin 0.1N KOH