Learn More
Bioworld Kinetin, 100 g
Supplier:  Bioworld 306310145
Encompass_Preferred
Kinetin (6-Furfuryl-aminopurine)Synonym: N6-FurfuryladenineCytokinin growth regulatorPlant tissue-culture testedAppearance: white crystaline powderMelting Point: 269-271°CSoluble in 1N NaOHSolubilizeat 1 mg/mLin 0.1N KOH
Catalog No. 50-255-059