Learn More
Selleck Chemical LLC Kinetin 100mg 525-79-1 6-Furfuryladenine
SDP
Catalog No. 508250
Encompass_Preferred
Kinetin (6-Furfuryladenine) is a type of cytokinin, a class of plant hormone that promotes cell division. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-825-0 Supplier Selleck Chemical LLC Supplier No. S2316100MG
Kinetin (6-Furfuryladenine) is a type of cytokinin, a class of plant hormone that promotes cell division. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.