Learn More
Alkali Scientific Kinetin [6 Furfurylaminopurine], 1 G
SDP
Supplier:  Alkali Scientific KIN001CP
Encompass_Preferred
Kinetin (6-Furfurylaminopurine) is a cytokinin widely used in plant tissue culture to promote cell division, shoot initiation, and overall plant growth. This 1g formulation is particularly effective in stimulating the development of lateral shoots and enhancing plant regeneration from cultured tissues. Kinetin plays a crucial role in delaying senescence and can influence the differentiation of plant cells into specific tissues. Its application is essential in tissue culture systems where rapid plant regeneration or vegetative propagation is required. Kinetin is also used in biotechnology for enhancing crop yields and improving resistance to stress by influencing plant growth patterns and metabolic pathways.
Catalog No. 50-006-59895