Learn More
Alkali Scientific Kinetin [6 Furfurylaminopurine], 5 G
SDP
Supplier:  Alkali Scientific KIN002CP
Encompass_Preferred
Kinetin (6-Furfurylaminopurine) is a potent cytokinin used in plant tissue culture to induce cell division, promote shoot formation, and enhance plant regeneration. This 5g formulation is ideal for large-scale tissue culture applications where high concentrations of kinetin are required for efficient plant propagation and growth. Kinetin has been shown to delay the senescence of plant tissues and improve the overall vigor of regenerated plants. It is essential for enhancing regeneration rates in various plant species, making it a crucial tool in agriculture, horticulture, and plant biotechnology for improving crop yields and plant stress resistance.
Catalog No. 50-006-59763