Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kinetin, >95%, MP Biomedicals™

Catalog No. ICN10211701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10211701 1 g
ICN10211705 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10211701 Supplier MP Biomedicals, Inc Supplier No. 0210211701
Add to Cart
Add to Cart

Chemical Identifiers

CAS 525-79-1
Molecular Formula C10H9N5O
Molecular Weight (g/mol) 215.216
InChI Key QANMHLXAZMSUEX-UHFFFAOYSA-N
Synonym kinetin, 6-furfurylaminopurine, 6-furfurylamino purine, 6-furfuryladenine, n6-furfuryladenine, n-furfuryladenine, cytokinin, n-furan-2-ylmethyl-9h-purin-6-amine, n6-furfurylamino purine, adenine, n-furfuryl
PubChem CID 3830
ChEBI CHEBI:27407
IUPAC Name N-(furan-2-ylmethyl)-7H-purin-6-amine
SMILES C1=COC(=C1)CNC2=NC=NC3=C2NC=N3

Specifications

Color White
Quantity 1 g
Formula Weight 215.2
Percent Purity >95%
Physical Form Powder
Chemical Name or Material Kinetin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.