Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Kinetin riboside | 4338-47-0 | MFCD00037987 | 99.9% | 347.33 g·mol⁻¹ | C15H17N5O5 | 1 ML
SDP

Catalog No. 5000212061
Encompass_Preferred

Kinetin riboside (N6-Furfuryladenosine) is a cytokinin analog reported to induce apoptosis in cancer cells and to inhibit HCT-15 proliferation with an IC50 of 2.5 μM. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL), it is intended for cell-based assays and biochemical research requiring a soluble cytokinin analog.

  • Ready-to-use 10 mM solution in DMSO
  • High purity (99.88%) suitable for research use
  • Reported to induce apoptosis and inhibit HCT-15 cells (IC50 2.5 μM)
  • Molecular weight 347.33 g·mol⁻¹; formula C15H17N5O5
  • Store in solvent at -80°C for long-term stability

Catalog No. 50-002-12061 Supplier Medchemexpress LLC Supplier No. HY10105510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kinetin riboside (N6-Furfuryladenosine) is a cytokinin analog reported to induce apoptosis in cancer cells and to inhibit HCT-15 proliferation with an IC50 of 2.5 μM. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL), it is intended for cell-based assays and biochemical research requiring a soluble cytokinin analog.

  • Ready-to-use 10 mM solution in DMSO
  • High purity (99.88%) suitable for research use
  • Reported to induce apoptosis and inhibit HCT-15 cells (IC50 2.5 μM)
  • Molecular weight 347.33 g·mol⁻¹; formula C15H17N5O5
  • Store in solvent at -80°C for long-term stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.