Learn More
Bioworld Kinetin Solution, 1mg/mL, 100 mL
Catalog No. 50255091
Encompass_Preferred
Kinetin (6-Furfurylaminopurine) solutionSpecification:Grade: Biotechnology GradeConcentration: 1mg/mlSterility: Sterile filtered
Catalog No. 50-255-091 Supplier Bioworld Supplier No. 306330021
Kinetin (6-Furfurylaminopurine) solutionSpecification:Grade: Biotechnology GradeConcentration: 1mg/mlSterility: Sterile filtered