Learn More
Medchemexpress LLC Kinetin triphosphate tetrasodium | 00-00-0 | 99.9% | C15H16N5Na4O14P3 | 10MG
SDP
Catalog No. 5000259052
Encompass_Preferred
Kinetin triphosphate tetrasodium is an ATP analogue used in biochemical and kinase research, including investigations of mitochondrial quality control pathways. It is supplied as a tetrasodium salt in small milligram quantities and includes documentation for experimental use and handling.
  • ATP analogue for kinase and enzymology assays.
  • High purity (99.92%) suitable for sensitive biochemical studies.
  • Soluble in water with recommended reconstitution guidance (100 mM preparations).
  • Provided in small research quantities for experimental flexibility.
  • Documentation includes datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-59052 Supplier Medchemexpress LLC Supplier No. HY134398A10MG
Kinetin triphosphate tetrasodium is an ATP analogue used in biochemical and kinase research, including investigations of mitochondrial quality control pathways. It is supplied as a tetrasodium salt in small milligram quantities and includes documentation for experimental use and handling.
  • ATP analogue for kinase and enzymology assays.
  • High purity (99.92%) suitable for sensitive biochemical studies.
  • Soluble in water with recommended reconstitution guidance (100 mM preparations).
  • Provided in small research quantities for experimental flexibility.
  • Documentation includes datasheet, certificate of analysis, and safety data sheet.