Learn More
Medchemexpress LLC Kinetin triphosphate tetrasodium | 99.9% | 675.2 g/mol | C15H16N5Na4O14P3 | 5 MG
SDP
Catalog No. 5000222053
Encompass_Preferred
Kinetin triphosphate tetrasodium is an ATP analogue provided as the tetrasodium salt for biochemical research. It modulates kinase activity and has documented use in studies such as Parkinson's disease research. Supplied as a small, concentrated vial suitable for in vitro kinase assays and mechanistic studies.
  • High purity (99.92%).
  • Supplied as the tetrasodium salt, water-soluble for aqueous assays.
  • Provided in small vial sizes (5 mg; 100 mM when dissolved as indicated by supplier).
  • Suitable for kinase activity assays and mechanistic studies.
  • Datasheet, certificate of analysis, and safety data sheet available for quality and safety information.
Catalog No. 50-002-22053 Supplier Medchemexpress LLC Supplier No. HY134398A5MG
Kinetin triphosphate tetrasodium is an ATP analogue provided as the tetrasodium salt for biochemical research. It modulates kinase activity and has documented use in studies such as Parkinson's disease research. Supplied as a small, concentrated vial suitable for in vitro kinase assays and mechanistic studies.
  • High purity (99.92%).
  • Supplied as the tetrasodium salt, water-soluble for aqueous assays.
  • Provided in small vial sizes (5 mg; 100 mM when dissolved as indicated by supplier).
  • Suitable for kinase activity assays and mechanistic studies.
  • Datasheet, certificate of analysis, and safety data sheet available for quality and safety information.