Learn More
Sigma Aldrich Fine Chemicals Biosciences King Agar B, 500 g
Catalog No. NC1073331
Encompass
For the detection and enumeration of fluorescing bacteria in water, in particular Pseudomonas fluorescens in drinking water.
Catalog No. NC1073331 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 60786500G
For the detection and enumeration of fluorescing bacteria in water, in particular Pseudomonas fluorescens in drinking water.