Learn More
Macherey-Nagel KingFisher Accessory Kit B
Catalog No. NC1729158
Encompass_Preferred
KingFisher Accessory Kit B for 4 x 96 of NucleoMag Blood 200 µL and NucleoMag 96 Plant / RNA preps using a KingFisher Flex platform - 20 KingFisher Deep-well Blocks, 4 KingFisher Deep-well Tip Combs, 4 KingFisher Elution Plates
Catalog No. NC1729158 Supplier Macherey-Nagel Supplier No. 744951
KingFisher Accessory Kit B for 4 x 96 of NucleoMag Blood 200 µL and NucleoMag 96 Plant / RNA preps using a KingFisher Flex platform - 20 KingFisher Deep-well Blocks, 4 KingFisher Deep-well Tip Combs, 4 KingFisher Elution Plates