Learn More
Description
The KingFisher™ Flex 24 Deep-Well Heating Block is compatible with the KingFisher™ Flex 24 Deep-Well Plate when used on the KingFisher™ Flex Magnetic Particle Processor with 24 Deep-Well Head.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Format | 24 Deep-well Heating Block |
| Capacity | 24 Samples |
| For Use With (Equipment) | KingFisher™ Flex |
| Product Line | KingFisher |
| Quantity | Each |
| Shipping Condition | Room Temperature |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
