Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ KingFisher™ Flex-Ready DNA Ultra 2.0 Prefilled Plates

Catalog No. FERA36586 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Reactions:
96 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
FERA36586 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERA36586 Supplier Thermo Scientific™ Supplier No. A36586
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Automation-ready plates prefilled with MagMAX DNA Multi-Sample Ultra 2.0 reagents suitably configured for KingFisher Flex purification systems

  • Enable isolation of genomic DNA from 50 to 400 μL of multiple sample types such as whole blood, saliva, buffy coat, buccal swabs, and more. The purified gDNA is ideal for a variety of downstream molecular biology applications
  • Key features of the KingFisher Flex-Ready DNA Ultra 2.0 Prefilled Plates:
    • Maximize convenience with MagMAX DNA Multi-Sample Ultra 2.0 isolation chemistry prefilled into KingFisher SBS microtiter plates
    • Increase ease of use—process 96 samples on instrument without the hassle of reagent setup
    • Designed to reduce user error
    • Helps to reduce hands-on time
    • Formatted specifically for KingFisher Flex instruments
    • All the benefits of MagMAX sample prep chemistries in automation-ready plate format
    • Eliminate the need to aliquot magnetic beads, lysis solutions, and wash solutions. Just peel off the foil seals from the KingFisher Flex-Ready DNA Ultra 2.0 prefilled plates; add enhancer, 50-400 μL of your sample, and proteinase K; and load all of the plates into the KingFisher Flex instrument
    • Eliminate the need for pipetting reagents, searching for plastics, or risking reagent mix-ups in your DNA isolation workflow
  • Complete, prefilled automation-ready sample prep kit improves convenience with all accessory plastics
  • What you get:
    • Seven foil-sealed KingFisher Flex-Ready DNA Ultra 2.0 Prefilled Plates sufficient to process 96 samples in one instrument run (96 samples in total)
    • One empty and sealed sample input 96 deep well plate
    • KingFisher 96-tip comb
    • Proteinase K solution and enhancer solution sufficient for 96 samples
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
For Use With (Equipment) KingFisher Flex purification systems
For Use With (Application) DNA extraction
No. of Reactions 96 Preps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.