Learn More
Thermo Scientific™ KingFisher Plastic Consumables for SARS-CoV-2 Testing
Shop All Thermo Scientific ProductsDescription
These KingFisher compatible plastic consumables are offered specifically for labs performing COVID-19 testing with the MagMAX Viral/Pathogen Nucleic Acid Purification Kit and the TaqPath COVID-19 Combo Kit.
For TaqPath COVID-19 testing with MagMAX Viral/Pathogen Nucleic Acid Isolation Kit
Specifications
Specifications
| Format | 96-well Plate |
| Capacity | 96 Samples |
| For Use With (Equipment) | KingFisher Flex and Presto |
| Includes | KingFisher DW 96 Tip Comb |
| Quantity | 100/Pk. |
Frequently Asked Questions (FAQs)
The alternative is KingFisher Plastics for 96 deep‐well format (Cat. No. 97002534). This product is a 96 tip comb for deep‐well magnets that can be used on the KingFisher Flex and Presto instruments.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.